Learn More
RETSCH Stainless Steel Grinding Jar
Product Code. 15370994 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
For CryoMill
For EMAX
For MM 200
Capacity (Metric):
1.5 mL
5 mL
10 mL
35 mL
50 mL
125 mL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Capacity (Metric) Unit Size
15370994 For CryoMill 35 mL Each
10504041 For MM 200 1.5 mL Each
14821631 For CryoMill 10 mL Each
10034170 For MM 200 10 mL Each
15300974 For EMAX 125 mL Each
15370964 For EMAX 50 mL Each
10399770 For MM 200 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15370994 Supplier RETSCH Supplier No. 01.462.0333
  • ONLINE EXCLUSIVE PRICE
    13910.00 NOK
    12651.75 NOK / Pack of 1
    Save 1258.25 NOK
    9% Off
Estimated Shipment: 20-01-2027
Use these grinding jars for dry, wet, and cryogenic grinding of small samples. Retsch™ Stainless Steel Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
  • Screw-top grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Product Type Grinding Jar
Capacity (Metric) 35 mL
For Use With (Equipment) For CryoMill
Material Stainless Steel