Learn More
RETSCH Stainless Steel Grinding Balls
Product Code. p-8007380 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
Planetary Ball Mill PM 400
Reaction Vials
Diameter (Metric):
3 mm
4 mm
5 mm
40 mm
Unit Size:
Each
Pack of 200
Product Code. For Use With (Equipment) Diameter (Metric) unitSize
10208591 Planetary Ball Mill PM 400 40 mm Each
10146641 Reaction Vials 5 mm Pack of 200
10196541 Reaction Vials 3 mm Each
11727664 Reaction Vials 4 mm Pack of 200
4 options
Product Code. 10208591
Brand: RETSCH 05.368.0060
Only null left
Add to basket
Add to basket
Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.
  • For use with MM 200 and MM 400 Mixer Mills
Specifications
Product Type Grinding Balls
Diameter (Metric) 40 mm
For Use With (Equipment) Planetary Ball Mill PM 400
Material Stainless Steel