Learn More
Cole-Parmer™ Stainless-steel Griffin-Style Beaker with Easy-Pour Rim
Product Code. 11772315 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (Metric) Outer:
66.6 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
11772315 66.6 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11772315 Supplier Cole-Parmer™ Supplier No. 0720540
  • ONLINE EXCLUSIVE PRICE
    797.00 NOK
    781.25 NOK / Pack of 1
    Save 15.75 NOK
    1% Off
View all promotions
Estimated Shipment: 29-12-2026
Assure accurate measuring with these beakers. Cole-Parmer™ Stainless-steel Griffin-Style Beaker with Easy-Pour Rim are great for higher temperatures and the general stirring or measuring of solutions.
  • Durable, economical mixing capabilities
  • Medium gauge 304 SS
  • Measure volumes easily with internal graduation markers
Specifications
Material Stainless Steel
Height (Metric) 80 mm
Product Type Beaker
Diameter (Metric) Outer 66.6 mm