Learn More
Cole-Parmer™ Stainless Steel Funnel
Product Code. 11777944 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
67 mL
70 mL
188 mL
350 mL
385 mL
658 mL
Height (Metric):
82.55 mm
104.1 mm
134.6 mm
160 mm
180.3 mm
210.8 mm
Unit Size:
Each
Product Code. Capacity (Metric) Height (Metric) unitSize
11777944 658 mL 160 mm Each
11761856 385 mL 134.6 mm Each
11787944 350 mL 180.3 mm Each
11797944 67 mL 210.8 mm Each
11741856 70 mL 82.55 mm Each
11751856 188 mL 104.1 mm Each
6 options
Product Code. 11777944
Brand: Cole-Parmer™ 0726600
Only null left
Add to basket
Add to basket
Stainless steel funnels are a laboratory supply you will count on for years of use.
  • Stay in stock with our selection of medium gauge, 304 stainless steel funnels
  • Available in various capacities and overall sizes
Specifications
Capacity (Metric) 658 mL
Material Stainless Steel
Height (Metric) 160 mm