Learn More
Cole-Parmer™ Stainless Steel Funnel
Product Code. 11761856 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
67 mL
70 mL
188 mL
350 mL
385 mL
658 mL
Height (Metric):
82.55 mm
104.1 mm
134.6 mm
160 mm
180.3 mm
210.8 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) unitSize
11761856 385 mL 134.6 mm Each
11777944 658 mL 160 mm Each
11787944 350 mL 180.3 mm Each
11797944 67 mL 210.8 mm Each
11741856 70 mL 82.55 mm Each
11751856 188 mL 104.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11761856 Supplier Cole-Parmer™ Supplier No. 0726500
  • ONLINE EXCLUSIVE PRICE
    1265.00 NOK
    1234.08 NOK / Pack of 1
    Save 30.92 NOK
    2% Off
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Stainless steel funnels are a laboratory supply you will count on for years of use.
  • Stay in stock with our selection of medium gauge, 304 stainless steel funnels
  • Available in various capacities and overall sizes
Specifications
Capacity (Metric) 385 mL
Material Stainless Steel
Height (Metric) 134.6 mm