Learn More
S Murray and Co™ Stainless Steel Forceps
Product Code. p-8000755
Change view
Click to view available options
Length (Metric):
110 mm
125 mm
Tip Style:
Spatulate
Straight
Unit Size:
Each
Product Code. Length (Metric) Tip Style unitSize
12730046 125 mm Straight Each
12342158 110 mm Spatulate Each
2 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12730046
Brand: S Murray and Co™ E051/01
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Use these forceps for a wide range of applications. S Murray™ Stainless Steel Forceps are made from only high grade stainless steel in accordance to the specifications of ISO 7153-1.
  • Stainless Steel forceps
  • For dissection applications
Specifications
Product Type Forceps
Material Stainless Steel
For Use With (Application) Dissection applications
Quantity Each
Tip Style Straight
Length (Metric) 125 mm