Learn More
S Murray and Co™ Stainless Steel Forceps
Product Code. 12730046
Change view
Click to view available options
Length (Metric):
110 mm
125 mm
Tip Style:
Spatulate
Straight
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style Unit Size
12730046 125 mm Straight Each
12342158 110 mm Spatulate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12730046 Supplier S Murray and Co™ Supplier No. E051/01
  • ONLINE EXCLUSIVE PRICE
    49.65 NOK
    27.82 NOK / Pack of 1
    Save 21.83 NOK
    43% Off
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Use these forceps for a wide range of applications. S Murray™ Stainless Steel Forceps are made from only high grade stainless steel in accordance to the specifications of ISO 7153-1.
  • Stainless Steel forceps
  • For dissection applications
Specifications
Product Type Forceps
Material Stainless Steel
For Use With (Application) Dissection applications
Quantity Each
Tip Style Straight
Length (Metric) 125 mm