Learn More
Cole-Parmer™ Stainless Steel Forcep
Product Code. 11705798 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
30.48 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11705798 30.48 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11705798 Supplier Cole-Parmer™ Supplier No. 0728810
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
Recommended for micro electronics and precision assembly
  • Finely crafted stainless steel tweezers
  • Feature hand finished points for accuracy and excellent tip hardness
  • Anti-glare finish reduces eye strain
  • Anti-magnetic, anti-acid and acid resistant
Specifications
Product Type Forceps
Length (English) 12 in.
Length (Metric) 30.48 cm
Material Stainless Steel
Sterility Sterile
Tip Style Rounded