Description
- Especially sturdy model
- Stainless steel AISI 304 (1.4301)
- With handy suspension outlet
- Seamless connection of scoop and hollow handle, therefore without crevices and edges
- Especially smooth, highly polished surface
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Product Type | Food Scoop |
| For Use With (Application) | Suitable for Use with Foodstuffs |
| Width (Metric) | 85 mm |
| Dimensions (L x W) | 155 x 85 mm |
| Material | Stainless Steel (AISI 304 (1.4301)) |
| Length (Metric) | 155 mm |