Learn More
Agencinox™ Stainless Steel Flat Bottom Dish
Product Code. 11847503
Change view
Click to view available options
Capacity (Metric):
20 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 5
Product Code. Capacity (Metric) unitSize
11847503 250 mL Each
11729503 20 mL Pack of 5
11776533 500 mL Each
11786533 1000 mL Each
4 options
Product Code. 11847503
Brand: Agencinox™ 310A
Only null left
Add to basket
Add to basket
Use these dishes in decontamination services, operating rooms, surgeries, clinics and healthcare fields. Agencinox™ Stainless Steel Flat Bottom Dishes are ideal for holding dressings and other items which may be needed during surgical procedures.
  • Flat bottom stainless steel lotion bowls
  • Autoclavable
  • Available in a variety of sizes
Specifications
Bottom Shape Flat
Capacity (Metric) 250 mL
Material Stainless Steel
Shape Round
Type Dish
Height (Metric) 50 mm
Diameter (Metric) 100 mm