Learn More
Agencinox™ Stainless Steel Flat Bottom Dish with Handle
Description
- Flat bottom stainless steel lotion bowls
- Autoclavable
- Available in a variety of sizes
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 500 mL |
| Material | Stainless Steel |
| Shape | Round |
| Type | Dish |
| Height (Metric) | 60 mm |
| Diameter (Metric) | 120 mm |