Learn More
Grant Instruments™ Stainless Steel Flask Clamp
Product Code. 15315852
Change view
Click to view available options
For Use With (Equipment):
1000mL Flask
100mL Flask
250mL Flask
25mL Flask
500mL Flask
50mL Flask
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15315852 25mL Flask Each
15325862 1000mL Flask Each
15365852 100mL Flask Each
15385852 250mL Flask Each
15305862 500mL Flask Each
15335852 50mL Flask Each
6 options
Product Code. 15315852
Brand: Grant Instruments™ SC25
Only null left
Add to basket
Add to basket
Holds flasks in place when used with Grant Aqua Pro Shaking Baths
  • Spring clamp provides security while allowing easy flask insertion/removal
  • For use with OLS and LSB Model Aqua Pro Shaking Water Baths
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 25mL Flask
Type Flask Clamp
Material Stainless Steel