Learn More
OHAUS™ Stainless Steel Flask Clamp
Product Code. 15792499 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
10 mL Flask
125 mL Flask
25 mL Flask
250 mL Flask
50 mL Flask
500 mL Flask
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15792499 125 mL Flask Each
15762499 10 mL Flask Each
15772499 25 mL Flask Each
15782499 50 mL Flask Each
15702509 250 mL Flask Each
15712509 500 mL Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15792499 Supplier OHAUS™ Supplier No. 30400087
Only null left
Add to basket
Add to basket
OHAUS™ Stainless Steel Flask Clamp is designed to securely grip and position various sizes of flasks.
  • Constructed of stainless steel
  • For use with flasks
  • Designed to securely hold flask in place
TRUSTED_SUSTAINABILITY
Specifications
Type Flask Clamp
Material Stainless Steel
For Use With (Equipment) 125 mL Flask
Description Flask Clamp, 125 mL
Color Silver
Jaw Type Four Interlocking