Learn More
OHAUS™ Stainless Steel Flask Clamp
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
10 mL Flask
125 mL Flask
25 mL Flask
250 mL Flask
50 mL Flask
500 mL Flask
Unit Size:
Each
Description
- Constructed of stainless steel
- For use with flasks
- Designed to securely hold flask in place
Specifications
Specifications
| Type | Flask Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | 25 mL Flask |
| Description | Flask Clamp, 25 mL |
| Color | Silver |
| Jaw Type | Four Interlocking |