Learn More
S Murray and Co™ Stainless Steel Featherweight Spring Forceps
Product Code. 12301379
Change view
Click to view available options
Length (Metric):
102 mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12301379 102 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12301379 Supplier S Murray and Co™ Supplier No. E015/01
  • ONLINE EXCLUSIVE PRICE
    233.00 NOK
    194.31 NOK / Pack of 5
    Save 38.69 NOK
    16% Off
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
Use these forceps for general laboratory applications. S Murray™ Stainless Steel Featherweight Spring Forceps are suitable for theatre, ward, veterinary, dental and chiropody use.
Use these forceps for general laboratory applications. S Murray™ Stainless Steel Featherweight Spring Forceps are suitable for theatre, ward, veterinary, dental and chiropody use.
  • Manufactured from high grade stainless steel
  • Complies with ISO 7153-1 specifications
  • Standard instruments are finished to a bright mirror polish
Specifications
Product Type Featherweight Spring Forceps
Material Stainless Steel
For Use With (Application) For dissection
Quantity 5/Pk.
Length (Metric) 102 mm