Learn More
- Home
- Products
- First Aid and Medical
- Patient Care Products
- Medical Syringes
- BD Microlance™ Stainless Steel Needles
BD Microlance™ Stainless Steel Needles
Product Code. 10595314| Product Code. | Needle Gauge | Length (English) Needle | Diameter (Metric) Outer | Color | unitSize |
|---|---|---|---|---|---|
| 10595314 | 22 G | 1.0 in. | 0.7 mm | Black | Pack of 100 |
| 10703815 | 26 G | 0.375 in. | 0.45 mm | Brown | Pack of 100 |
| 10442014 | 30 G | 0.5 in. | 0.29 mm | Yellow | Pack of 100 |
| 10753785 | 26 G | 0.5 in. | 0.45 mm | Brown | Pack of 100 |
| 10204444 | 27 G | 0.5 in. | 0.4 mm | Gray | Pack of 100 |
| 12349169 | 26 G | 0.625 in. | 0.45 mm | Brown | Pack of 100 |
| 10442204 | 25 G | 0.625 in. | 0.5 mm | Orange | Pack of 100 |
| 12329179 | 21 G | 0.625 in. | 0.8 mm | Green | Pack of 100 |
| 12389169 | 25 G | 1.0 in. | 0.5 mm | Orange | Pack of 100 |
| 12399169 | 23 G | 1.0 in. | 0.6 mm | Blue | Pack of 100 |
| 10030464 | 20 G | 1.0 in. | 0.9 mm | Yellow | Pack of 100 |
| 12309179 | 23 G | 1.25 in. | 0.6 mm | Blue | Pack of 100 |
| 12379179 | 19 G | 1.0 in. | 1.1 mm | Beige | Pack of 100 |
| 10472204 | 21 G | 1.5 in. | 0.8 mm | Green | Pack of 100 |
| 10201211 | 20 G | 1.5 in. | 0.9 mm | Yellow | Pack of 100 |
| 10234154 | 19 G | 1.5 in. | 0.9 mm | Beige | Pack of 100 |
| 10518554 | 16 G | 1.5 in. | 1.6 mm | White | Pack of 100 |
| 10775295 | 21 G | 2.0 in. | 0.8 mm | Green | Pack of 100 |
| 10162534 | 18 G | 1.5 in. | 1.25 mm | Pink | Pack of 100 |
| 12369179 | 19 G | 2.0 in. | 1.1 mm | Beige | Pack of 100 |
| 10020654 | 18 G | 2.0 in. | 1.25 mm | Pink | Pack of 100 |
Product Code. 10595314
Brand: BD 304727
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Unique slip angle and polishing method results in less patient discomfort at the injection site and less risk for punching.
- Hypodermic needles with thin walls for optimal flow
- Fits syringes connected to a Luer adapter or Luer Lock connector
- Color coded for quick recognition of gauge for easy selection of appropriate flow rate
- Sterile and individually wrapped for hygienic, single-use application
- Stainless steel needles with epoxy resin adhesives
Specifications
Specifications
| Material | Stainless Steel with Epoxy Resin Adhesives |
| Color | Black |
| Description | 5,000 individually wrapped needles, normal cannula wall |
| Needle Gauge | 22 G |
| Diameter (Metric) Outer | 0.7 mm |
| For Use With (Application) | For single use only |
| Latex Free | Yes |
| Length (English) Needle | 1.0 in. |
| Product Type | Needle |
| Needle Point Style | Bevel |
| For Use With (Equipment) | SZR -150, connected to a Luer adapter or Luer Lock connector |
| Sterility | Sterile |
| Show More Show Less |