Learn More
Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors
Click to view available options
Length (Metric):
130 mm
140 mm
155 mm
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
Unit Size:
Each
Description
- Three types: round-round, sharp-round, and sharp-sharp
- Stainless steel
Specifications
Specifications
| Product Type | Dressing Scissors |
| Length (Metric) | 155 mm |
| Material | Stainless Steel |
| Tip Style | Sharp/Sharp |