Learn More
Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors
Product Code. 10208930
Change view
Click to view available options
Length (Metric):
130 mm
140 mm
155 mm
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
Unit Size:
Each
Product Code. Length (Metric) Tip Style unitSize
10208930 140 mm Sharp/Round Each
10540042 130 mm Round/Round Each
10088360 130 mm Sharp/Sharp Each
10231961 155 mm Round/Round Each
10733521 140 mm Round/Round Each
10787191 155 mm Sharp/Round Each
10777191 155 mm Sharp/Sharp Each
7 options
Product Code. 10208930
Brand: Bochem™ 4101
Only null left
Add to basket
Add to basket
Manufactured from stainless steel. Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors comes in three types.
  • Three types: round-round, sharp-round, and sharp-sharp
  • Stainless steel
Specifications
Product Type Dressing Scissors
Length (Metric) 140 mm
Material Stainless Steel
Tip Style Sharp/Round