Learn More
Schneider Gerd™ Stainless Steel Double Micro-spatula
Product Code. 10787085
Change view
Click to view available options
Length (Metric):
210 mm
Width (Metric):
6 mm
Unit Size:
Each
Product Code. Length (Metric) Width (Metric) unitSize
10787085 210 mm 6 mm Each
1 options
Product Code. 10787085
Brand: Schneider Gerd™ 364
Only null left
Add to basket
Add to basket
Use this micro spatula for a variety of applications. Schneider Gerd™ Stainless Steel Double Micro-spatulas are ideal for use with a wide range of compounds.
  • Spatulas are ideal for working with powders, stirring and mixing
  • Perfect for working with miniscule amounts of chemicals
  • Two different end shapes, offer increased versatility
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Quantity 1 Ea.
Shape Double
Product Type Double Ended Micro Spatula
Width (Metric) 6 mm
Material Stainless Steel
Length (Metric) 210 mm