Learn More
Nickel Electro™ Stainless Steel Dissection Dish
Shop All Nickel-Electro Products
Click to view available options
Dimensions (L x W x H):
200 x 310 x 45 mm
300 x 450 x 50 mm
360 x 260 x 50 mm
Unit Size:
Each
Description
- Stainless steel
- Shallow form
- One piece pressing
- Useful when working with and transporting small specimens
Dissecting dish, stainless steel, one piece pressing
Specifications
Specifications
| Bottom Shape | Flat |
| Material | Stainless Steel |
| Shape | Rectangular |
| Type | Dissection Dish |
| Dimensions (L x W x H) | 360 x 260 x 50 mm |
| Length (Metric) | 360 mm |
| Width (Metric) | 260 mm |
| Height (Metric) | 50 mm |