Learn More
Schneider Gerd™ Stainless Steel Dissecting Needle
Product Code. 10525544
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 10525544
Brand: Schneider Gerd™ 642
With plastic handle
  • Stainless Steel Dissecting Needle
  • 150 x 1.0mm
  • Plastic handle
Specifications
Material Stainless Steel
Color Black Handle
Description 150 x 1.0mm
For Use With (Application) Dissecting
Product Type Dissecting Needle