Description
- Self-closing lid with damping, large, ergonomically shaped handle on the rear of the container for easy emptying of the container
- Opening by large pedal with anti-slip surface
- FM-certified
Specifications
Specifications
| Capacity (Metric) | 50 L |
| Depth (English) | 19.49 in. |
| Depth (Metric) | 495.00 mm |
| Capacity (English) | 13.2 gal. |
| Color | Gray |
| Height (Metric) | 540.00 mm |
| Weight (English) | 18.74 lb. |
| Weight (Metric) | 8.5 kg |
| Width (Metric) | 505.00 mm |
| Width (English) | 19.88 in. |
| Show More |