Learn More
KGW Isotherm™ Stainless Steel Dewar Flasks
Specifications
Specifications
| Height (Metric) | 312mm |
| Capacity (Metric) | 2 L |
| Diameter (Metric) Outer | 122 mm |
| Material | Stainless Steel |