Description
- Well-suited for pharmaceutical and environmental sampling
- Ideal for weighing and handling of samples
- Great for weighing messy ingredients, tiny objects and portions
- Great accessory to have if you own a weighing scale
- Perfect for weighing anything from liquid to gems and other materials
Specifications
Specifications
| Shape | Cylindrical |
| Product Type | Scoop |
| Width (Metric) | 30 mm |
| Dimensions (L x W) | 90 x 30 mm |
| Material | Stainless Steel |
| Length (Metric) | 90 mm |