Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class E2 with Certificate
Shop All Fisherbrand Products
Click to view available options
Weight (Metric):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
200 g
Unit Size:
1 unit
Description
- Class E2 stainless steel calibration weight
- In compliance with OIML R111
- Delivered with a calibration certificate guaranteeing traceability
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Classification | E2 |
| Weight (Metric) | 1 g to 5 kg |
| Mass | 1 g to 5 kg |