Learn More
AMSTA Rostfritt Stainless Steel Container with Lid
Product Code. 11723839
Change view
Click to view available options
Capacity (Metric):
1 L
3 L
5 L
10 L
20 L
25 L
30 L
Includes:
Lid
Lid and 3 Clamps
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes unitSize
11723839 25 L Lid Each
15750429 10 L Lid Each
11762074 1 L Lid and 3 Clamps Each
11805650 3 L Lid Each
15760429 5 L Lid Each
11752074 5 L Lid and 3 Clamps Each
11722074 20 L Lid and 3 Clamps Each
11742074 30 L Lid and 3 Clamps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11723839 Supplier AMSTA Rostfritt Supplier No. 51608251
High-quality stainless steel container, available in various sizes with or without clamps and tap.
  • Quality
  • Invisible weld seams
  • Pharmaceutical grade finish
  • Sustainable production
  • Made in Sweden
Specifications
Product Type Can
Capacity (Metric) 25 L
Color Silver
Material Stainless Steel (316L)
Finish Ra 0.2 to 0.3 μm
Includes Lid
Height (Metric) 397 mm
Diameter (Metric) Outer 300 mm