Learn More
Bochem™ Stainless Steel Container
Product Code. 10761962
Change view
Click to view available options
Diameter (Metric) Inner:
270mm
300mm
400mm
450mm
Capacity (Metric):
20 L
25 L
75 L
100 L
Height (Metric):
375mm
600mm
670mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Capacity (Metric) Height (Metric) Unit Size
10761962 300mm 25 L 375mm Each
10342711 270mm 20 L 375mm Each
10020811 400mm 75 L 600mm Each
10172791 450mm 100 L 670mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10761962 Supplier Bochem™ Supplier No. 8341
Estimated Shipment: 01-02-2027
Bochem™ Stainless Steel Container is constructed of Stainless Steel and is equipped with handles on the side.
  • Stainless Steel
  • Includes handles
  • Compatible lid sold separately
Specifications
Diameter (Metric) Inner 300mm
Height (Metric) 375mm
Capacity (Metric) 25 L
Material Stainless Steel (18/10)