Learn More
Bochem™ Stainless Steel Container
Product Code. 10342711
Change view
Click to view available options
Diameter (Metric) Inner:
270mm
300mm
400mm
450mm
Capacity (Metric):
20 L
25 L
75 L
100 L
Height (Metric):
375mm
600mm
670mm
Unit Size:
Each
Product Code. Diameter (Metric) Inner Capacity (Metric) Height (Metric) unitSize
10342711 270mm 20 L 375mm Each
10020811 400mm 75 L 600mm Each
10761962 300mm 25 L 375mm Each
10172791 450mm 100 L 670mm Each
4 options
Product Code. 10342711
Brand: Bochem™ 8340
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bochem™ Stainless Steel Container is constructed of Stainless Steel and is equipped with handles on the side.
  • Stainless Steel
  • Includes handles
  • Compatible lid sold separately
Specifications
Diameter (Metric) Inner 270mm
Height (Metric) 375mm
Capacity (Metric) 20 L
Material Stainless Steel (18/10)