missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Stainless Steel Comfort Racks for Chest Freezers

With spring locks mounted on each shelf to secure cryoboxes. Tenak™ Stainless Steel Comfort Racks for Chest Freezers can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate.

Brand:  Tenak™ TE24464

Additional Details : Weight : 1.80000kg

 View more versions of this product

Product Code. 15280794

  • 1210.00 NOK / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • Available in a range of heights
  • Can be used with all freezer models
  • Sold without cryoboxes
Specifications

Specifications

Spring Lock
Boxes (14)
Chest freezer
140 x 140 x 504mm
504 mm
140 mm
Comfort Rack
Stainless Steel
1 Pack
140 mm
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers