Learn More
Tenak™ Stainless Steel Comfort Rack for Upright Freezers
Product Code. 15105952 Shop All TENAK Products
Change view
Click to view available options
Holds:
Boxes (2 x 5/10)
Boxes (28)
Boxes (3 x 3/9)
Boxes (3 x 4/12)
Boxes (4 x 3/12)
Boxes (4 x 4/16)
Boxes (5 x 3/15)
Boxes (5 x 4/20)
Boxes (5 x 5/25)
Microtiterplates (3 x 5/15)
Microtiterplates (3 x 6/18)
Microtiterplates (4 x 5/20)
Microtiterplates (5 x 5/25)
Height (Metric):
200 mm
214 mm
228 mm
246 mm
261 mm
266 mm
285 mm
332 mm
398 mm
Unit Size:
Each
Product Code. Holds Height (Metric) unitSize
15105952 Boxes (4 x 3/12) 228 mm Each
15240734 Boxes (5 x 3/15) 285 mm Each
15220754 Boxes (5 x 5/25) 285 mm Each
12497060 Microtiterplates (5 x 5/25) 332 mm Each
15160875 Boxes (4 x 4/16) 228 mm Each
12477060 Microtiterplates (3 x 5/15) 200 mm Each
12487060 Microtiterplates (4 x 5/20) 266 mm Each
12487080 Microtiterplates (3 x 6/18) 200 mm Each
15210744 Boxes (5 x 4/20) 285 mm Each
15240764 Boxes (3 x 4/12) 246 mm Each
15240784 Boxes (2 x 5/10) 214 mm Each
15280754 Boxes (3 x 3/9) 246 mm Each
15290784 Boxes (3 x 3/9) 261 mm Each
15250744 Boxes (28) 398 mm Each
14 options
Product Code. 15105952
Brand: Tenak™ TE24222
Only null left
Add to basket
Add to basket
Place the cryoboxes in drawers with the Tenak™ Stainless Steel Comfort Rack for Upright Freezers .This comfort rack structure increases the flexibility and reduces the operating weight when compared to rack types.
  • Six different box heights
  • Fit all freezer models
  • Stainless steel.
  • Cryoboxes sold separately
Specifications
Cell Dividers Yes
Holds Boxes (4 x 3/12)
Material Stainless Steel
Positions 12
For Use With (Equipment) Upright Freezers
No. of Racks 12
No. per Pack 1 Pack
Quantity 1 Pack
Depth (Metric) 421 mm
Product Type Comfort Rack
Height (Metric) 228 mm
Array 4 x 3
Width (Metric) 140 mm