Learn More
Nickel Electro™ Stainless Steel Flask Clip
Product Code. 12778968 Shop All Nickel-Electro Products
Change view
Click to view available options
Holds:
1 L Conical Flask
100 mL Conical Flask
2 L Conical Flask
2.8 L Fernbach flask
25 mL Conical Flask
250 mL Conical Flask
50 mL Conical Flask
500 mL Conical Flask
Unit Size:
Each
Product Code. Holds unitSize
12778968 2.8 L Fernbach flask Each
11479629 1 L Conical Flask Each
11776577 2 L Conical Flask Each
11736577 25 mL Conical Flask Each
11766577 250 mL Conical Flask Each
11469629 500 mL Conical Flask Each
11746577 50 mL Conical Flask Each
11756577 100 mL Conical Flask Each
8 options
Product Code. 12778968
Brand: Nickel Electro™ FC3000/FB
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For personalised clip arrangement, clips are supplied with all necessary fixings to enable them to be attached to the Perforated Tray. Nickel Electro™ Stainless Steel Flask Clip is suitable for flasks and can be autoclaved.
  • Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
  • No sharp edges to cut or score glassware
  • Can be autoclaved
  • Plastic base
  • Easy to assemble and remove Flask Clips from Perforated tray
Specifications
Holds 2.8 L Fernbach flask
Product Type Flask Clip
Material Stainless Steel