Learn More
Nickel Electro™ Stainless Steel Flask Clip
Product Code. 11766577 Shop All Nickel-Electro Products
Change view
Click to view available options
Holds:
1 L Conical Flask
100 mL Conical Flask
2 L Conical Flask
2.8 L Fernbach flask
25 mL Conical Flask
250 mL Conical Flask
50 mL Conical Flask
500 mL Conical Flask
Unit Size:
Each
Product Code. Holds unitSize
11766577 250 mL Conical Flask Each
11479629 1 L Conical Flask Each
11776577 2 L Conical Flask Each
11736577 25 mL Conical Flask Each
11469629 500 mL Conical Flask Each
11746577 50 mL Conical Flask Each
11756577 100 mL Conical Flask Each
12778968 2.8 L Fernbach flask Each
8 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11766577
Brand: Nickel Electro™ FC0250
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
For personalised clip arrangement, clips are supplied with all necessary fixings to enable them to be attached to the Perforated Tray. Nickel Electro™ Stainless Steel Flask Clip is suitable for flasks and can be autoclaved.
  • Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
  • No sharp edges to cut or score glassware
  • Can be autoclaved
  • Plastic base
  • Easy to assemble and remove Flask Clips from Perforated tray
Specifikationer
Holds 250 mL Conical Flask
Product Type Flask Clip
Material Stainless Steel
For Use With (Equipment) 250 mL Conical Flask
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.