Learn More
Nickel Electro™ Stainless Steel Flask Clip
Shop All Nickel-Electro Products
Click to view available options
Holds:
1 L Conical Flask
100 mL Conical Flask
2 L Conical Flask
2.8 L Fernbach flask
25 mL Conical Flask
250 mL Conical Flask
50 mL Conical Flask
500 mL Conical Flask
Unit Size:
Each
Description
- Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
- No sharp edges to cut or score glassware
- Can be autoclaved
- Plastic base
- Easy to assemble and remove Flask Clips from Perforated tray
Specifications
Specifications
| Holds | 100 mL Conical Flask |
| Product Type | Flask Clip |
| Material | Stainless Steel |
| Includes | Perforated mixing disc |