Description
- Fits standard sized boxes with 9x9 and 10x10 cell dividers
- Max. footprint of 134x134 mm
- Rack for 50mm box height
Specifications
Specifications
| Cell Dividers | Yes |
| Holds | 12 |
| Material | Stainless Steel |
| Positions | 12 |
| For Use With (Application) | Secure the Cryoboxes |
| For Use With (Equipment) | Cryogenic freezer |
| No. per Pack | 1 Pack |
| Quantity | 1 Pack |
| Size | For Box Height: 50 mm |
| Depth (Metric) | 140 mm |
| Show More |