Learn More
Bochem™ Stainless Steel Clamp
Product Code. 10088630
Click to view available options
Diameter (Metric):
60 mm
Length (Metric) Arm:
150 mm
Jaw Type:
Round
Unit Size:
Each
Product Code. 10088630
Brand: Bochem™ 5403
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel. BOCHEM™ Stainless Steel Clamps hold test tubes and other equipment most often used in titration experiments.
  • 18/10 stainless steel
  • Round jaws
  • DIN 12894
Specifications
Quantity 1 Ea.
Diameter (Metric) 60 mm
Length (Metric) Arm 150 mm
Jaw Type Round