Learn More
RETSCH Stainless Steel Cassette Pan
Brand:  RETSCH 03.010.0037
2850.00 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.50000kg
Product Code. 11914282
  • 3000.00 NOK / Each
Estimated Shipment: 21-08-2024
Specifications
Cassette Pan
900 mL
For Ultra Centrifugal Mill ZM 200