Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F2, NVLAP
Product Code. 16682252 Shop All Ohaus Products
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
Unit Size:
Each
Product Code. Mass unitSize
16682252 2 g Each
16682262 1 mg Each
16672262 2 mg Each
16662262 5 mg Each
16652262 10 mg Each
16642262 20 mg Each
16632262 50 mg Each
16622262 100 mg Each
16612262 200 mg Each
16602262 500 mg Each
16692252 1 g Each
16672252 5 g Each
16662252 10 g Each
16652252 20 g Each
16642252 50 g Each
16632252 100 g Each
16622252 200 g Each
16612252 500 g Each
16552592 1 kg Each
16602252 2 kg Each
16692242 5 kg Each
16682242 10 kg Each
16672242 20 kg Each
23 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16682252
Marke: Ohaus™ 30390890
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F2 Tolerances
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Spezifikation
Certifications/Compliance NVLAP Accredited Certificate
Material Troemner Alloy 8
Mass 2 g
Tolerance 0.400 mg
Includes Protective Case