Learn More
Amsta Rostfritt Stainless Steel Bottle - Acid Proof Without Lid, With Skirtband
Product Code. p-200113412
Click to view available options
Capacity (Metric):
4 L
5 L
6 L
8 L
10 L
Unit Size:
Each
Pack of 10
Product Code. 30285057
Brand: Amsta Rostfritt 51603100
Highest quality stainless steel bottle - acid proof. Made of 1 mm stainless steel (EN1.4404). Intended for antibiotics, vaccines, chemicals and liquids. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Product Type Pail
Capacity (Metric) 10 L
Color Silver
Material Stainless Steel (316L)