Learn More
- Home
- Products
- Beakers and Lids
- Beakers
- Bochem™ Stainless Steel Beakers
Bochem™ Stainless Steel Beakers
Product Code. 10142651
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Includes:
Rim
Rim and Spout
Rim, Handle
Rim, Spout
Rim, Spout, Handle
Unit Size:
Each
| Product Code. | Capacity (Metric) | Includes | unitSize |
|---|---|---|---|
| 10142651 | 250 mL | Rim, Handle | Each |
| 10586181 | 100 mL | Rim, Handle | Each |
| 10781392 | 100 mL | Rim, Spout, Handle | Each |
| 10498871 | 250 mL | Rim, Spout, Handle | Each |
| 10201081 | 250 mL | Rim | Each |
| 10663891 | 250 mL | Rim, Spout | Each |
| 10288690 | 500 mL | Rim, Spout, Handle | Each |
| 10017730 | 500 mL | Rim | Each |
| 10603511 | 1000 mL | Rim | Each |
| 10470061 | 1000 mL | Rim, Handle | Each |
| 10218840 | 1000 mL | Rim, Spout, Handle | Each |
| 10343191 | 1000 mL | Rim and Spout | Each |
| 10566932 | 2000 mL | Rim, Handle | Each |
| 10303051 | 2000 mL | Rim | Each |
| 10627341 | 2000 mL | Rim, Spout, Handle | Each |
| 10330731 | 3000 mL | Rim, Spout, Handle | Each |
| 10363091 | 3000 mL | Rim | Each |
| 10271361 | 3000 mL | Rim, Spout | Each |
| 10729621 | 5000 mL | Rim | Each |
| 10453501 | 5000 mL | Rim, Spout | Each |
| 10244081 | 5000 mL | Rim, Handle | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- 18/10 stainless steel construction.
- Resistant and durable.
- Contact bottom for heating plates and magnetic stirrers.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Stainless Steel (18/10) |
| Includes | Rim, Handle |
| Type | Beaker |