Learn More
Stainless Steel, Alloy 254SMO, S31254, ARMI∣MBH
Description
Stainless Steel, Alloy 254SMO, S31254, Ø40x3mm
Specifications
Specifications
| Chemical Name or Material | Niobium/Tantalum/Zirconium/Boron/Sulfur/Antimony/Calcium/Oxygen/Titanium/Tin/Arsenic/Carbon/Aluminium/Phosphorus/Tungsten/Vanadium/Cobalt/Nitrogen/Silicon/Copper/Manganese/Molybdenum/Nickel/Chromium/Iron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Niobium -83 ppm/Tantalum -46 ppm/Zirconium -17 ppm/Boron 0.0009 %/Sulfur 0.001 %/Antimony 0.0017 %/Calcium 0.0024 %/Oxygen 0.0025 %/Titanium 0.004 %/Tin 0.0041 %/Arsenic 0.0044 %/Carbon 0.017 %/Aluminium 0.022 %/Phosphorus 0.022 %/Tungsten 0.041 %/Vanadium 0.052 %/Cobalt 0.118 %/Nitrogen 0.2 %/Silicon 0.301 %/Copper 0.77 %/Manganese 0.892 %/Molybdenum 6.09 %/Nickel 18.1 %/Chromium 19.7 %/Iron 53.7 % |