Learn More
Hamilton™ Stainless Steel Anion Exhchange Columns
Product Code. 11503504
Click to view available options
Length (Metric):
50 mm
250 mm
Particle Size:
7 μm
10 μm
Anion Exchange Capacity:
0.11 μeq/g
0.19 μeq/g
1.6 μeq/g
35 μeq/g
Unit Size:
Each
Product Code. 11503504
Brand: Hamilton™ 79365
Specifications
Anion Exchange Capacity 0.19 μeq/g
Description PRP-X100
Particle Size 10 μm
Stationary Phase PRP-X100
Material Stainless Steel
Packing Material PSDVB/Trimethylammonium
Pore Size 100 &‌Aring;
Length (Metric) 50 mm
Diameter (Metric) Inner 4.1 mm
Diameter (Metric) 4.1 mm