Learn More
Hamilton™ Stainless Steel Anion Exhchange Columns
Product Code. 10573923
Change view
Click to view available options
Length (Metric):
50 mm
250 mm
Particle Size:
7 μm
10 μm
Anion Exchange Capacity:
0.11 μeq/g
0.19 μeq/g
1.6 μeq/g
35 μeq/g
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Particle Size Anion Exchange Capacity unitSize
10573923 250 mm 7 μm 1.6 μeq/g Each
11503504 50 mm 10 μm 0.19 μeq/g Each
10452543 250 mm 7 μm 0.11 μeq/g Each
11583504 - 10 μm 35 μeq/g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10573923 Supplier Hamilton™ Supplier No. 79828
Estimated Shipment: 10-09-2026
Specifications
Anion Exchange Capacity 1.6 μeq/g
Description PRP-X800
Column Capacity 2.56 meq/column
Particle Size 7 μm
Stationary Phase PRP-X800
Material Stainless Steel
Packing Material PSDVB/Itaconate
Pore Size 100 &‌Aring;
Temperature (Metric) 5°C to 60°C
Length (Metric) 250 mm
Diameter (Metric) Inner 4.1 mm
Diameter (Metric) 4.1 mm