Learn More
Hamilton™ Stainless Steel Anion Exhchange Columns
Product Code. 10452543
Click to view available options
Length (Metric):
50 mm
250 mm
Particle Size:
7 μm
10 μm
Anion Exchange Capacity:
0.11 μeq/g
0.19 μeq/g
1.6 μeq/g
35 μeq/g
Unit Size:
Each
Product Code. 10452543
Brand: Hamilton™ 79734
Specifications
Anion Exchange Capacity 0.11 μeq/g
Description PRP-X110
Column Capacity 0.176 meq/column
Particle Size 7 μm
Stationary Phase PRP-X110
Material Stainless Steel
Packing Material PSDVB/Trimethylammonium
Pore Size 100 &‌Aring;
Temperature (Metric) 5°C to 60°C
USP Type L47
Length (Metric) 250 mm
Diameter (Metric) Inner 4.1 mm
Diameter (Metric) 4.1 mm