Learn More
Julabo™ Stainless Steel 1-Adaptor
Product Code. 11910288
Change view
Click to view available options
Connection Type:
M16 x 1 Male to 1/2 in. BSP Female
M16 x 1 Male to 3/4 in. BSP Female
Unit Size:
Each
Product Code. Connection Type unitSize
11910288 M16 x 1 Male to 3/4 in. BSP Female Each
10752604 M16 x 1 Male to 1/2 in. BSP Female Each
2 options
Product Code. 11910288
Brand: Julabo™ 8891009
Only null left
Specifications
For Use With (Equipment) Suitable for CF30, CF31, CF40, CF41 Cryo-Compact Circulator
Type 1-Adapter
Material Stainless Steel
Connection Type M16 x 1 Male to 3/4 in. BSP Female