Learn More
Metro™ Stainless Lab Worktable, Stainless Island Top and 3-Sided Frame
Keep lab materials clean and safe. Metro Stainless Lab Worktable features Type 304 stainless steel construction throughout and 3-sided Frame providing corrosion-proof performance and stability you can count on.
Brand:  Metro™ LTS48UIS
Additional Details : Weight : 32.48000kg
 View more versions of this product
Product Code. 15907111
  • 21420.00 NOK / Each
Estimated Shipment: 04-07-2024
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
47.75 in.
47.75 x 30 x 35 in. (121.2 x 76.2 x 89 cm)
89 cm
3 Sided frame
273 kg
121.2 cm
Stainless Steel
Stationary Table
Ideal for lab
35 in.
30 in.
76.2 cm
600 lb.