Learn More
B Braun™ Hypodermic Needles
Product Code. 10482792
Click to view available options
Needle Gauge:
18 G
20 G
21 G
23 G
26 G
27 G
Length (Metric) Needle:
20 mm
25 mm
30 mm
40 mm
50 mm
80 mm
120 mm
Diameter (Metric) Outer:
0.4 mm
0.45 mm
0.6 mm
0.8 mm
0.9 mm
1.2 mm
Color:
Blue
Brown
Gray
Green
Pink
Yellow
Unit Size:
Pack of 100
Product Code. 10482792
Brand: B Braun™ 4657640
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for patient comfort in a broad array of sizes. B Braun™ Hypodermic Needles provide a smooth surface with light silicone coating designed to minimize discomfort.
 Unique, highly automated grinding and polishing process and low-friction lubrication is designed to achieve unequaled sharpness and ease of penetration. Transparent hub design provides high visibility to detect flashback.
  • Color-coded hubs conform to ISO 6009 for easy identity
  • Wide range of diameters and lengths
  • Gauges from 18G to 27G
  • Ultra-sharp, tri-beveled stainless steel designed to result in minimal pain
Specifications
Material Polypropylene (Hub), Stainless Chromium Nickel Steel
Color Blue
Needle Gauge 23 G
Diameter (Metric) Outer 0.6 mm
For Use With (Application) For minimal pain upon puncture, single use
Latex Free Yes
Product Type Needle
Length (Metric) Needle 30 mm
Needle Point Style Ultra-sharp Long Tri-bevel