Learn More
B Braun™ Hypodermic Needles
Description
Unique, highly automated grinding and polishing process and low-friction lubrication is designed to achieve unequaled sharpness and ease of penetration. Transparent hub design provides high visibility to detect flashback.
- Color-coded hubs conform to ISO 6009 for easy identity
- Wide range of diameters and lengths
- Gauges from 18G to 27G
- Ultra-sharp, tri-beveled stainless steel designed to result in minimal pain
Specifications
Specifications
| Material | Polypropylene (Hub), Stainless Chromium Nickel Steel |
| Color | Pink |
| Needle Gauge | 18 G |
| Diameter (Metric) Outer | 1.2 mm |
| For Use With (Application) | For minimal pain upon puncture, single use |
| Latex Free | Yes |
| Product Type | Needle |
| Length (Metric) Needle | 40 mm |
| Needle Point Style | Ultra-sharp Long Tri-bevel |
| Sterility | Sterile |