Learn More
Grant Instruments™ Stainles Steel Clamp for Flasks
Product Code. p-7241266
Change view
Click to view available options
Holds:
100 mL flask
1000 mL flask
250 mL flask
50 mL flask
500 mL flask
Unit Size:
Each
Product Code. Holds unitSize
17795009 100 mL flask Each
17705019 1000 mL flask Each
17715019 250 mL flask Each
17725019 50 mL flask Each
17735019 500 mL flask Each
5 options
Product Code. 17795009
Brand: Grant Instruments™ HSC100
Only null left
Add to basket
Add to basket
For use with Grant Instruments UP-168 platform that fits in ES-20/80
Clamp, stainless steel,flask, UP-168, platform,ES-20/80, Grant Instruments
TRUSTED_SUSTAINABILITY
Specifications
Holds 100 mL flask
Product Type Clamp
Description Tight fit clamp 100 mL - 65 mm dia. for UP-168
For Use With (Equipment) UP-168