Learn More
Brady™ Stainerbondz Super Chemical Resistant Polyester Laboratory Labels
Click to view available options
Quantity:
2500/Pk.
Height (English):
0.75 in.
0.9 in.
Unit Size:
Each
Description
- StainerBondz™ B-481 matte white polyester has a permanent acrylic adhesive and is thermal transfer printable with B30 series printers from Brady™
- B-481 performs well in the commonly used H and E (hematoxilin and eosin) slide staining process
- Excellent resistance to a wide variety of chemicals, oils and water
- Service temperature of -112 to 266° F (-80 to 130° C)
Specifications
Specifications
| Certifications/Compliance | RoHS |
| Adhesive Type | Acrylic, Permanent |
| Coating | None |
| Color | White |
| Finish | Matte |
| Format | Die-cut |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | None |
| Product Type | Laboratory Label |
| Show More |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido