Learn More
Brady™ StainerBondz™ Microscope Slide Matte Polyester Laboratory Labels for M6/M7 Printers
Product Code. 17838284
Click to view available options
Dimensions (LxW):
0.375 x 0.9 in.
0.75 x 0.9 in.
0.9 x 0.9 in.
Height (Metric):
9.53 mm
19.05 mm
22.86 mm
Unit Size:
Pack of 250
Pack of 500
Product Code. 17838284
Brand: Brady™ 174088
Only null left
Add to basket
Add to basket
Super chemical-resistant polyester is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
  • Performs well in the commonly used H&E (hematoxilin and eosin) slide staining process
  • High resistance to a wide variety of chemicals, oils and water
  • Service temperature of -80°C to +130°C (-112°F to +266°F)
  • Made of B-481 polyester material
Specifications
Autoclavable Autoclavable
Certifications/Compliance ASTM D 1000, ASTM D 2979
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Healthcare and Hospital Labeling, General Identification, Laboratory Labeling
For Use With (Equipment) M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Latex Free No
Quantity 250 Labels
Shape Rectangle
Temperature Range (English) -112°F to +266°F
Temperature Range (Metric) -80°C to +130°C
Thickness (English) 0.0033 in.
Thickness (Metric) 0.084 mm
Dimensions (LxW) 0.75 x 0.9 in.
Length (English) 0.75 in.
Width (English) 0.9 in.
Width (Metric) 22.86 mm
Height (Metric) 19.05 mm
Alternate Part Number M6-194-481